Assay Details
Target Gene Details
Entrez Gene ID: | 54776 |
Gene Name: | protein phosphatase 1 regulatory subunit 12C |
Gene Aliases: |
LENG3, MBS85, p84, p85 |
Location: |
Chr.19:55090913-55117600 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R12C | NM_001271618.1 | NP_001258547.1 | ||
| NM_017607.3 | NP_060077.1 | |||
| XM_005259013.3 | XP_005259070.1 | |||
| XM_011527045.2 | XP_011525347.1 | |||
| AB209452.1 | BAD92689.1 | |||
| AF312028.1 | AAG60045.1 | |||
| AK027086.1 | BAB15651.1 | |||
| AK129529.1 | BAC85179.1 | |||
| AK297187.1 | ||||
| AK297426.1 | ||||
| AL137618.1 | CAB70844.1 | |||
| BC008030.1 | AAH08030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv817874 | Chr.19:55067334 - 55103555 on Build GRCh38 | Loss |
|
| esv2667609 | Chr.19:54472514 - 55094226 on Build GRCh38 | Deletion |
|
| esv3644829 | Chr.19:55030893 - 55138732 on Build GRCh38 | Gain |
|
| esv3644836 | Chr.19:55084362 - 55113486 on Build GRCh38 | Gain |
|
| esv3893251 | Chr.19:55079303 - 55094733 on Build GRCh38 | Loss |
|
| esv34356 | Chr.19:55026094 - 55139319 on Build GRCh38 | Gain |
|
| esv2718872 | Chr.19:54472594 - 55094247 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115703186] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map