Assay Details
Target Gene Details
Entrez Gene ID: | 92104 |
Gene Name: | tetratricopeptide repeat domain 30A |
Gene Aliases: |
IFT70A |
Location: |
Chr.2:177614298-177618966 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC30A | NM_152275.3 | 1 | 4103 | NP_689488.3 |
| AL049329.1 | 1 | 2204 | CAB43676.2 | |
| BM684657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583689 | Chr.2:177603280 - 177645092 on Build GRCh38 | Loss |
|
| esv2762964 | Chr.2:177551666 - 177617090 on Build GRCh38 | Loss |
|
| nsv583688 | Chr.2:177562912 - 177675565 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map