Assay Details
Target Gene Details
Entrez Gene ID: | 257194 |
Gene Name: | neuronal growth regulator 1 |
Gene Aliases: |
DMML2433, IGLON4, KILON, Ntra |
Location: |
Chr.1:71402942-72282850 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEGR1 | NM_173808.2 | NP_776169.2 | ||
| BC036771.1 | AAH36771.1 | |||
| BX538014.1 | CAD97961.1 | |||
| BX648027.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830159 | Chr.1:71306625 - 71444177 on Build GRCh38 | Gain |
|
| nsv830170 | Chr.1:71390764 - 71543075 on Build GRCh38 | Gain |
|
| dgv19n27 | Chr.1:71383354 - 71472011 on Build GRCh38 | Gain |
|
| nsv546474 | Chr.1:71327538 - 71574671 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map