Assay Details
Target Gene Details
Entrez Gene ID: | 84449 |
Gene Name: | zinc finger protein 333 |
Gene Aliases: |
- |
Location: |
Chr.19:14689787-14733746 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF333 | NM_001300912.1 | NP_001287841.1 | ||
| NM_032433.2 | 12 | 1175 | NP_115809.1 | |
| XM_005260098.3 | 13 | 1488 | XP_005260155.1 | |
| XM_006722924.2 | 12 | 1252 | XP_006722987.1 | |
| XM_011528362.2 | 12 | 1216 | XP_011526664.1 | |
| XM_011528363.2 | 14 | 2316 | XP_011526665.1 | |
| XM_011528365.2 | 14 | 1657 | XP_011526667.1 | |
| XM_011528366.2 | 13 | 1485 | XP_011526668.1 | |
| XM_011528367.2 | 13 | 1498 | XP_011526669.1 | |
| XM_011528368.2 | 13 | 1405 | XP_011526670.1 | |
| XM_011528369.2 | 13 | 1418 | XP_011526671.1 | |
| XM_011528371.2 | 6 | 1582 | XP_011526673.1 | |
| XM_011528372.2 | 5 | 511 | XP_011526674.1 | |
| XM_017027367.1 | 12 | 1192 | XP_016882856.1 | |
| XM_017027368.1 | 11 | 1060 | XP_016882857.1 | |
| XM_017027369.1 | 13 | 2813 | XP_016882858.1 | |
| XM_017027370.1 | 13 | 2826 | XP_016882859.1 | |
| XM_017027371.1 | 13 | 1438 | XP_016882860.1 | |
| AB058709.1 | 5 | 489 | BAB47435.1 | |
| AF372702.1 | 12 | 1132 | AAM15528.1 | |
| AK094028.1 | 13 | 1418 | ||
| AK095142.1 | 1 | 57 | ||
| AK122817.1 | 5 | 1056 | ||
| AL832544.1 | 6 | 667 | ||
| BC048107.1 | AAH48107.1 | |||
| BX647475.1 | 12 | 1136 | CAI56712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578680 | Chr.19:14695350 - 14735523 on Build GRCh38 | Loss |
|
| nsv1060306 | Chr.19:14697771 - 14721224 on Build GRCh38 | Loss |
|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map