Assay Details
Target Gene Details
Entrez Gene ID: | 8681 |
Gene Name: | JMJD7-PLA2G4B readthrough |
Gene Aliases: |
HsT16992, cPLA2-beta |
Location: |
Chr.15:41828085-41848148 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JMJD7-PLA2G4B | NM_001198588.1 | 19 | 2006 | NP_001185517.1 |
| NM_005090.3 | 19 | 2006 | NP_005081.1 | |
| AB209150.1 | 19 | 5849 | BAD92387.1 | |
| AF065215.1 | 19 | 1981 | AAC78836.1 | |
| AF065216.1 | 20 | 5409 | ||
| AF121908.1 | 19 | 1973 | AAD32135.1 | |
| DQ523799.1 | 19 | 1974 | ||
| DQ523800.1 | 19 | 1974 |
Target Gene Details
Entrez Gene ID: | 100137049 |
Gene Name: | phospholipase A2 group IVB |
Gene Aliases: |
HsT16992, cPLA2-beta |
Location: |
Chr.15:41838813-41848148 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G4B | NM_001114633.1 | 14 | 1374 | NP_001108105.1 |
| AK124489.1 | 6 | 1685 | ||
| AK299419.1 | 14 | 1340 | ||
| BF372565.1 | 4 | 440 | ||
| BX648318.1 | 3 | 2352 | ||
| CA488226.1 | 6 | 621 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain |
|
| nsv9244 | Chr.15:41818514 - 41916607 on Build GRCh38 | Gain |
|
| nsv1050892 | Chr.15:41836657 - 41894284 on Build GRCh38 | Gain |
|
| nsv1506 | Chr.15:41824733 - 41857173 on Build GRCh38 | Insertion |
|
| nsv952613 | Chr.15:41816803 - 41884902 on Build GRCh38 | Deletion |
|
| nsv517631 | Chr.15:41804728 - 41895294 on Build GRCh38 | Loss |
|
| dgv2582n100 | Chr.15:41791376 - 41900562 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map