Assay Details
Target Gene Details
Entrez Gene ID: | 81873 |
Gene Name: | actin related protein 2/3 complex subunit 5 like |
Gene Aliases: |
ARC16-2 |
Location: |
Chr.9:124862118-124877733 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPC5L | NM_030978.2 | 6 | 2162 | NP_112240.1 |
| XM_011519067.1 | 4 | 1296 | XP_011517369.1 | |
| AA731112.1 | 1 | 370 | ||
| AF087842.1 | AAP97155.1 | |||
| AK023881.1 | 1 | 1919 | ||
| AK056174.1 | 6 | 2150 | ||
| BC000798.1 | AAH00798.1 | |||
| BX537659.1 |
More Information
Set Membership: |
|

Genomic Map