Assay Details
Target Gene Details
Entrez Gene ID: | 4605 |
Gene Name: | MYB proto-oncogene like 2 |
Gene Aliases: |
B-MYB, BMYB |
Location: |
Chr.20:43667019-43716496 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYBL2 | NM_001278610.1 | NP_001265539.1 | ||
| NM_002466.3 | NP_002457.1 | |||
| AK223482.1 | BAD97202.1 | |||
| AK303249.1 | ||||
| AK314791.1 | ||||
| BC007585.1 | AAH07585.1 | |||
| BC053555.1 | AAH53555.1 | |||
| X13293.1 | CAA31655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645889 | Chr.20:43686024 - 43756429 on Build GRCh38 | Gain |
|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| esv3645886 | Chr.20:43653014 - 43756886 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map