Assay Details
Target Gene Details
Entrez Gene ID: | 2778 |
Gene Name: | GNAS complex locus |
Gene Aliases: |
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI |
Location: |
Chr.20:58839681-58911196 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAS | NM_016592.3 | 1 | 1084 | NP_057676.1 |
| XM_017027815.1 | XP_016883304.1 | |||
| XM_017027821.1 | 1 | 1121 | XP_016883310.1 | |
| XM_017027822.1 | 1 | 1121 | XP_016883311.1 | |
| AF105253.1 | 1 | 1062 | AAF63226.1 | |
| AK122771.1 | ||||
| AK314549.1 |
Target Gene Details
Entrez Gene ID: | 149775 |
Gene Name: | GNAS antisense RNA 1 |
Gene Aliases: |
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG |
Location: |
Chr.20:58818918-58850903 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAS-AS1 | NR_002785.2 | |||
| AJ251759.1 | ||||
| BF589977.1 | ||||
| DB067618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828763 | Chr.20:58839788 - 58908828 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map