Assay Details
Target Gene Details
Entrez Gene ID: | 114548 |
Gene Name: | NLR family pyrin domain containing 3 |
Gene Aliases: |
AGTAVPRL, AII, AVP, C1orf7, CIAS1, CLR1.1, FCAS, FCAS1, FCU, MWS, NALP3, PYPAF1 |
Location: |
Chr.1:247416156-247449108 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP3 | NM_001079821.2 | 5 | 1356 | NP_001073289.1 |
| NM_001127461.2 | 4 | 1998 | NP_001120933.1 | |
| NM_001127462.2 | 3 | 1964 | NP_001120934.1 | |
| NM_001243133.1 | 3 | 1964 | NP_001230062.1 | |
| NM_004895.4 | 3 | 1964 | NP_004886.3 | |
| NM_183395.2 | 3 | 1964 | NP_899632.1 | |
| XM_011544048.2 | 4 | 2103 | XP_011542350.1 | |
| XM_011544053.2 | 4 | 1991 | XP_011542355.1 | |
| XM_011544055.2 | 4 | 1991 | XP_011542357.1 | |
| XM_017000181.1 | 5 | 1580 | XP_016855670.1 | |
| XM_017000182.1 | 5 | 1525 | XP_016855671.1 | |
| XM_017000183.1 | 4 | 1991 | XP_016855672.1 | |
| XM_017000184.1 | 4 | 1991 | XP_016855673.1 | |
| AB208891.1 | 3 | 1973 | BAD92128.1 | |
| AF054176.1 | 1 | 43 | AAC39910.1 | |
| AF410477.1 | 3 | 1964 | AAL33908.1 | |
| AF418985.2 | 3 | 1380 | AAL14640.2 | |
| AF420469.1 | 4 | 1350 | AAL65136.1 | |
| AF427617.1 | 3 | 1964 | AAL33911.1 | |
| AF468522.1 | 3 | 1380 | AAL78632.1 | |
| AK027194.1 | 3 | 1393 | ||
| AK314998.1 | 5 | 1356 | ||
| AY092033.1 | 3 | 1380 | AAM14669.1 | |
| AY422168.1 | 3 | 1212 | AAQ98889.1 | |
| BC117211.1 | 3 | 1283 | ||
| BC143359.1 | 3 | 1283 | ||
| BC143362.1 | 3 | 1283 | ||
| BC143363.1 | 3 | 1283 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833137 | Chr.1:247274015 - 247444126 on Build GRCh38 | Gain |
|
| esv33129 | Chr.1:246911128 - 248165533 on Build GRCh38 | Gain+Loss |
|
| nsv1011097 | Chr.1:247402897 - 247503209 on Build GRCh38 | Gain |
|
| nsv509790 | Chr.1:247311000 - 247445876 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map