Assay Details
Target Gene Details
Entrez Gene ID: | 4709 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B3 |
Gene Aliases: |
B12, CI-B12 |
Location: |
Chr.2:201071739-201085750 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFB3 | NM_001257102.1 | NP_001244031.1 | ||
| NM_002491.2 | NP_002482.1 | |||
| XM_011511230.2 | XP_011509532.1 | |||
| XM_017004186.1 | XP_016859675.1 | |||
| AF035839.1 | AAC15590.1 | |||
| AF047183.1 | AAC04268.1 | |||
| BC018183.1 | AAH18183.1 | |||
| BU188560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584160 | Chr.2:201039216 - 201298760 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map