Assay Details
Target Gene Details
Entrez Gene ID: | 100532724 |
Gene Name: | NPHP3-ACAD11 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.3:132558138-132722459 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPHP3-ACAD11 | NR_037804.1 | |||
| AB082531.1 | BAC02709.1 | |||
| AY257867.1 | AAP83426.1 |
Target Gene Details
Entrez Gene ID: | 79876 |
Gene Name: | ubiquitin like modifier activating enzyme 5 |
Gene Aliases: |
THIFP1, UBE1DC1 |
Location: |
Chr.3:132654446-132679786 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA5 | NM_001320210.1 | 12 | 2259 | NP_001307139.1 |
| NM_001321238.1 | 10 | 2123 | NP_001308167.1 | |
| NM_001321239.1 | 10 | 2973 | NP_001308168.1 | |
| NM_024818.4 | 12 | 3109 | NP_079094.1 | |
| NM_198329.3 | 12 | 2132 | NP_938143.1 | |
| AL832669.1 | 8 | 3495 | ||
| AY253672.1 | 12 | 2261 | AAP79600.1 | |
| BU739244.1 | 1 | 417 | ||
| BX537742.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591798 | Chr.3:132626804 - 132975557 on Build GRCh38 | Loss |
|
| nsv591799 | Chr.3:132662755 - 133080365 on Build GRCh38 | Gain |
|
| nsv998211 | Chr.3:132605119 - 132988974 on Build GRCh38 | Loss |
|
| nsv4014 | Chr.3:132668918 - 132704197 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map