Assay Details
Target Gene Details
Entrez Gene ID: | 1368 |
Gene Name: | carboxypeptidase M |
Gene Aliases: |
- |
Location: |
Chr.12:68842200-68963240 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPM | NM_001005502.2 | 9 | 3591 | NP_001005502.1 |
| NM_001874.4 | 9 | 3623 | NP_001865.1 | |
| NM_198320.3 | 9 | 3609 | NP_938079.1 | |
| BG677445.1 | 1 | 269 | ||
| BQ431065.1 | 1 | 614 | ||
| CB993834.1 | 1 | 224 |
More Information
Set Membership: |
|

Genomic Map