Assay Details
Target Gene Details
Entrez Gene ID: | 25853 |
Gene Name: | DDB1 and CUL4 associated factor 12 |
Gene Aliases: |
CT102, KIAA1892, TCC52, WDR40A |
Location: |
Chr.9:34086383-34126773 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF12 | NM_015397.3 | 9 | 3586 | NP_056212.1 |
| XM_011517843.1 | 9 | 3413 | XP_011516145.1 | |
| AB067479.1 | 9 | 3506 | ||
| AF131771.1 | 4 | 2446 | ||
| AI140359.1 | 1 | 103 | ||
| AK000354.1 | 9 | 3364 | BAA91106.1 | |
| AL117646.1 | 6 | 2720 | CAB56028.2 | |
| BC008893.2 | 8 | 3149 | AAH08893.2 | |
| BC063823.1 | 9 | 3504 | AAH63823.1 | |
| BM144957.1 | 1 | 99 | ||
| BM194397.1 | 1 | 98 | ||
| DQ518909.1 | 9 | 3506 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain |
|
| nsv520467 | Chr.9:33804815 - 34146647 on Build GRCh38 | Gain |
|
| nsv6525 | Chr.9:34047777 - 34090442 on Build GRCh38 | Deletion |
|
| nsv1023129 | Chr.9:33967101 - 34118050 on Build GRCh38 | Gain |
|
| nsv1030732 | Chr.9:33973326 - 34314588 on Build GRCh38 | Gain |
|
| nsv614154 | Chr.9:34023301 - 34115078 on Build GRCh38 | Loss |
|
| esv3620345 | Chr.9:34080825 - 34134465 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map