Assay Details
Target Gene Details
Entrez Gene ID: | 4801 |
Gene Name: | nuclear transcription factor Y subunit beta |
Gene Aliases: |
CBF-A, CBF-B, HAP3, NF-YB |
Location: |
Chr.12:104117080-104138262 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFYB | NM_006166.3 | 8 | 2163 | NP_006157.1 |
| XM_005268908.4 | 9 | 2227 | XP_005268965.1 | |
| XM_005268909.4 | 9 | 2210 | XP_005268966.1 | |
| XM_005268910.4 | 9 | 2882 | XP_005268967.1 | |
| AU151875.1 | 1 | 309 |
More Information
Set Membership: |
|

Genomic Map