Assay Details
Target Gene Details
Entrez Gene ID: | 4953 |
Gene Name: | ornithine decarboxylase 1 |
Gene Aliases: |
ODC |
Location: |
Chr.2:10440371-10448554 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ODC1 | NM_001287188.1 | 12 | 1903 | NP_001274117.1 |
| NM_001287189.1 | 12 | 1895 | NP_001274118.1 | |
| NM_001287190.1 | 12 | 1741 | NP_001274119.1 | |
| NM_002539.2 | 12 | 2003 | NP_002530.1 | |
| AI277727.1 | 1 | 305 | ||
| AK292352.1 | 12 | 1876 | ||
| AK301956.1 | 13 | 1648 | ||
| BC025296.1 | 12 | 1743 | AAH25296.1 | |
| M16650.2 | 11 | 1529 | AAA59966.2 | |
| M20372.1 | 8 | 1110 | AAA59968.1 | |
| X55362.1 | 12 | 1744 | CAA39047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007185 | Chr.2:10400260 - 10651617 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map