Assay Details
Target Gene Details
Entrez Gene ID: | 148014 |
Gene Name: | tetratricopeptide repeat domain 9B |
Gene Aliases: |
- |
Location: |
Chr.19:40216058-40218694 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC9B | NM_152479.5 | NP_689692.2 | ||
| XM_017026368.1 | XP_016881857.1 | |||
| AK054935.1 | 1 | 424 | BAB70830.1 | |
| AK289550.1 | ||||
| BC029539.1 | AAH29539.1 | |||
| CA432622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644345 | Chr.19:40203124 - 40288712 on Build GRCh38 | Gain |
|
| esv2758761 | Chr.19:40106513 - 40333613 on Build GRCh38 | Gain |
|
| esv3644344 | Chr.19:40191059 - 40328585 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map