Assay Details
Target Gene Details
Entrez Gene ID: | 11009 |
Gene Name: | interleukin 24 |
Gene Aliases: |
C49A, FISP, IL10B, MDA7, MOB5, ST16 |
Location: |
Chr.1:206897404-206904139 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL24 | NM_001185156.1 | 7 | 877 | NP_001172085.1 |
| NM_001185157.1 | 6 | 718 | NP_001172086.1 | |
| NM_001185158.1 | 5 | 519 | NP_001172087.1 | |
| NM_006850.3 | 7 | 874 | NP_006841.1 | |
| XM_011509101.2 | 6 | 769 | XP_011507403.1 | |
| XM_017000121.1 | 6 | 767 | XP_016855610.1 | |
| AW949784.1 | 5 | 509 | ||
| AY641440.1 | 6 | 677 | AAV52800.1 | |
| AY641441.1 | 6 | 717 | AAV52801.1 | |
| BC009681.1 | 7 | 850 | AAH09681.1 | |
| BP222651.1 | 5 | 517 | ||
| BT007156.1 | AAP35820.1 | |||
| U16261.1 | 7 | 873 | AAA91780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010486 | Chr.1:206876486 - 207031685 on Build GRCh38 | Gain |
|
| nsv468049 | Chr.1:206894495 - 206963993 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map