Assay Details
Target Gene Details
Entrez Gene ID: | 27 |
Gene Name: | ABL proto-oncogene 2, non-receptor tyrosine kinase |
Gene Aliases: |
ABLL, ARG |
Location: |
Chr.1:179099327-179229693 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABL2 | NM_001136000.2 | NP_001129472.1 | ||
NM_001168236.1 | 11 | 2322 | NP_001161708.1 | |
NM_001168237.1 | NP_001161709.1 | |||
NM_001168238.1 | NP_001161710.1 | |||
NM_001168239.1 | NP_001161711.1 | |||
NM_005158.4 | 12 | 2098 | NP_005149.4 | |
NM_007314.3 | 12 | 2385 | NP_009298.1 | |
XM_005245088.2 | 11 | 2020 | XP_005245145.1 | |
XM_017001035.1 | 11 | 4695 | XP_016856524.1 | |
BC065912.1 | 12 | 2065 | AAH65912.1 | |
BX538317.1 | CAD98092.1 | |||
FJ542283.1 | ||||
FJ542284.1 | ||||
FJ542285.1 | 11 | 2035 | ||
FJ542286.1 | ||||
M35296.1 | 12 | 2302 | AAA35553.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002511 | Chr.1:179108779 - 179120707 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
