Assay Details
Target Gene Details
Entrez Gene ID: | 55247 |
Gene Name: | nei like DNA glycosylase 3 |
Gene Aliases: |
FGP2, FPG2, NEI3, ZGRF3, hFPG2, hNEI3 |
Location: |
Chr.4:177309837-177362938 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEIL3 | NM_018248.2 | 6 | 896 | NP_060718.2 |
| XM_017008360.1 | 6 | 862 | XP_016863849.1 | |
| AB079071.1 | 6 | 779 | BAC06479.1 | |
| AK001720.1 | 6 | 859 | BAA91860.1 | |
| AK307692.1 | ||||
| BC025954.1 | 6 | 852 | AAH25954.1 | |
| BE865481.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3603300 | Chr.4:177297546 - 177343289 on Build GRCh38 | Gain |
|
| esv2759304 | Chr.4:177244315 - 177484220 on Build GRCh38 | Gain |
|
| nsv822852 | Chr.4:177295992 - 177380771 on Build GRCh38 | Gain |
|
| esv34846 | Chr.4:177307859 - 177379956 on Build GRCh38 | Gain |
|
| esv3569839 | Chr.4:177338047 - 177341549 on Build GRCh38 | Loss |
|
| esv2728772 | Chr.4:175817087 - 177875422 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map