Assay Details
Target Gene Details
Entrez Gene ID: | 51170 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 11 |
Gene Aliases: |
17-BETA-HSD11, 17-BETA-HSDXI, 17BHSD11, DHRS8, PAN1B, RETSDR2, SDR16C2 |
Location: |
Chr.4:87336522-87391303 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSD17B11 | NM_016245.4 | 2 | 481 | NP_057329.3 |
| XM_011532021.1 | 2 | 430 | XP_011530323.1 | |
| AF126780.1 | 2 | 436 | AAF06939.1 | |
| AF273056.1 | 2 | 334 | AAM44459.1 | |
| AK075348.1 | 2 | 365 | BAC11560.1 | |
| AY358553.1 | 2 | 348 | AAQ88917.1 | |
| BC008650.1 | 2 | 371 | AAH08650.1 | |
| BC014327.1 | 2 | 347 | AAH14327.1 | |
| BC016367.1 | 2 | 378 | AAH16367.1 | |
| BC021673.1 | 2 | 352 | AAH21673.1 | |
| BC036001.1 | 2 | 372 | AAH36001.1 | |
| DA265697.1 | 2 | 480 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004518 | Chr.4:87297321 - 87433251 on Build GRCh38 | Gain |
|
| esv3601271 | Chr.4:87289406 - 87409657 on Build GRCh38 | Loss |
|
| esv3601274 | Chr.4:87364198 - 87422760 on Build GRCh38 | Loss |
|
| nsv1002789 | Chr.4:87318188 - 87685333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map