Assay Details
Target Gene Details
Entrez Gene ID: | 11234 |
Gene Name: | HPS5, biogenesis of lysosomal organelles complex 2 subunit 2 |
Gene Aliases: |
AIBP63, BLOC2S2 |
Location: |
Chr.11:18278670-18322498 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPS5 | NM_007216.3 | 10 | 1398 | NP_009147.3 |
| NM_181507.1 | 11 | 1555 | NP_852608.1 | |
| NM_181508.1 | 10 | 1211 | NP_852609.1 | |
| XM_011519862.1 | 11 | 1546 | XP_011518164.1 | |
| XM_011519863.1 | 11 | 1454 | XP_011518165.1 | |
| XM_011519864.1 | 11 | 1386 | XP_011518166.1 | |
| XM_011519865.1 | 10 | 1768 | XP_011518167.1 | |
| XM_011519866.1 | 10 | 1419 | XP_011518168.1 | |
| XM_011519867.1 | 10 | 1285 | XP_011518169.1 | |
| XM_011519868.2 | 10 | 1526 | XP_011518170.1 | |
| XM_017017149.1 | 11 | 1454 | XP_016872638.1 | |
| XM_017017150.1 | 11 | 1386 | XP_016872639.1 | |
| XM_017017151.1 | 10 | 1604 | XP_016872640.1 | |
| XM_017017152.1 | 10 | 1768 | XP_016872641.1 | |
| XM_017017153.1 | 10 | 1604 | XP_016872642.1 | |
| XM_017017154.1 | 10 | 1285 | XP_016872643.1 | |
| AB023234.1 | 10 | 1211 | ||
| AF534400.1 | 10 | 1211 | AAO25962.1 | |
| AF534401.1 | 11 | 1555 | AAO25963.1 | |
| AF534402.1 | 10 | 1398 | AAO25964.1 | |
| AK291663.1 | 11 | 1521 | ||
| AK292436.1 | 10 | 1364 | ||
| BC033640.1 | 10 | 1359 | AAH33640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map