Assay Details
Target Gene Details
Entrez Gene ID: | 25792 |
Gene Name: | CDKN1A interacting zinc finger protein 1 |
Gene Aliases: |
LSFR1, NP94, ZNF356 |
Location: |
Chr.9:128166065-128204383 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621804 | Chr.9:128166528 - 128179690 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map