Assay Details
Target Gene Details
Entrez Gene ID: | 5653 |
Gene Name: | kallikrein related peptidase 6 |
Gene Aliases: |
Bssp, Klk7, PRSS18, PRSS9, SP59, hK6 |
Location: |
Chr.19:50958631-50969673 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK6 | NM_001319948.1 | 1 | 111 | NP_001306877.1 |
| NM_002774.3 | 1 | 111 | NP_002765.1 | |
| AF013988.1 | AAB66483.1 | |||
| AK314897.1 | ||||
| AY279383.1 | 1 | 112 | ||
| D78203.1 | BAA11306.1 | |||
| U62801.1 | 1 | 112 | AAB07113.1 |
More Information
Set Membership: |
|

Genomic Map