Assay Details
Target Gene Details
Entrez Gene ID: | 128869 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class U |
Gene Aliases: |
CDC91L1, GAB1 |
Location: |
Chr.20:34560542-34677285 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGU | NM_080476.4 | NP_536724.1 | ||
| XM_011528542.2 | XP_011526844.1 | |||
| XM_017027664.1 | XP_016883153.1 | |||
| AB086842.1 | BAC53626.1 | |||
| AK075507.1 | BAC11660.1 | |||
| AK301900.1 | ||||
| AK308627.1 | ||||
| AY339061.1 | AAQ18022.1 | |||
| AY358816.1 | AAQ89175.1 | |||
| AY422169.1 | AAR23798.1 | |||
| BC030512.1 | AAH30512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3360 | Chr.20:34562312 - 34597446 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map