Assay Details
Target Gene Details
Entrez Gene ID: | 6011 |
Gene Name: | G protein-coupled receptor kinase 1 |
Gene Aliases: |
GPRK1, RHOK, RK |
Location: |
Chr.13:113645771-113735664 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK1 | NM_002929.2 | 2 | 856 | NP_002920.1 |
| XM_017020684.1 | 2 | 2523 | XP_016876173.1 | |
| AK289912.1 | 2 | 980 | ||
| BC140945.1 | 2 | 821 | ||
| L77503.1 | 2 | 763 | AAG50439.1 | |
| U63973.1 | 2 | 856 | AAB05929.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116539667] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map