Assay Details
Target Gene Details
Entrez Gene ID: | 171389 |
Gene Name: | NLR family pyrin domain containing 6 |
Gene Aliases: |
AVR, CLR11.4, NALP6, NAVR, NAVR/AVR, PAN3, PYPAF5 |
Location: |
Chr.11:278365-285942 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP6 | NM_001276700.1 | 4 | 1141 | NP_001263629.1 |
| NM_138329.2 | 4 | 1141 | NP_612202.2 | |
| XM_011519931.1 | 4 | 1346 | XP_011518233.1 | |
| XM_017017252.1 | 4 | 1346 | XP_016872741.1 | |
| XM_017017253.1 | 1 | 825 | XP_016872742.1 | |
| AF479748.1 | 4 | 1141 | AAL87105.1 | |
| AK292668.1 | 4 | 1346 | ||
| AY154461.1 | 4 | 1141 | AAO18157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32593 | Chr.11:218394 - 285937 on Build GRCh38 | Loss |
|
| esv3891905 | Chr.11:269147 - 336332 on Build GRCh38 | Gain |
|
| nsv552765 | Chr.11:199256 - 320836 on Build GRCh38 | Gain |
|
| dgv186n67 | Chr.11:280568 - 281471 on Build GRCh38 | Gain |
|
| dgv1505n54 | Chr.11:198510 - 470331 on Build GRCh38 | Gain |
|
| nsv482153 | Chr.11:278570 - 285303 on Build GRCh38 | Gain |
|
| nsv552766 | Chr.11:248181 - 285936 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map