Assay Details
Target Gene Details
Entrez Gene ID: | 7080 |
Gene Name: | NK2 homeobox 1 |
Gene Aliases: |
BCH, BHC, NK-2, NKX2.1, NKX2A, NMTC1, T/EBP, TEBP, TITF1, TTF-1, TTF1 |
Location: |
Chr.14:36516397-36520225 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX2-1 | NM_001079668.2 | 3 | 2035 | NP_001073136.1 |
| NM_003317.3 | 2 | 2190 | NP_003308.1 | |
| BC006221.2 | 3 | 1969 | AAH06221.2 | |
| BM970287.1 | 1 | 163 | ||
| U33749.1 | 2 | 2190 | AAB52381.1 | |
| U43203.1 | 3 | 2021 | AAA89066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819045 | Chr.14:36516524 - 36518288 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map