Assay Details
Target Gene Details
Entrez Gene ID: | 5294 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma |
Gene Aliases: |
PI3CG, PI3K, PI3Kgamma, PIK3, p110gamma, p120-PI3K |
Location: |
Chr.7:106865278-106908978 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CG | NM_001282426.1 | 11 | 4609 | NP_001269355.1 |
| NM_001282427.1 | 11 | 4533 | NP_001269356.1 | |
| NM_002649.3 | 11 | 4758 | NP_002640.2 | |
| XM_005250443.3 | 11 | 4626 | XP_005250500.1 | |
| BX648341.1 | 11 | 4591 | ||
| X83368.1 | 11 | 4768 | CAA58284.1 |
More Information
Set Membership: |
|

Genomic Map