Assay Details
Target Gene Details
Entrez Gene ID: | 51650 |
Gene Name: | mitochondrial ribosomal protein S33 |
Gene Aliases: |
CGI-139, MRP-S33, PTD003, S33mt |
Location: |
Chr.7:141006161-141014981 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS33 | NM_016071.3 | NP_057155.1 | ||
| NM_053035.2 | NP_444263.1 | |||
| AF078858.1 | AAD44490.1 | |||
| AF151897.1 | AAD34134.1 | |||
| AK024776.1 | ||||
| BC015462.2 | AAH15462.1 | |||
| BC105793.1 | AAI05794.1 | |||
| BP870829.1 | ||||
| BQ420818.1 | ||||
| W95986.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6645n100 | Chr.7:140902063 - 141046545 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map