Assay Details
Target Gene Details
Entrez Gene ID: | 55055 |
Gene Name: | zwilch kinetochore protein |
Gene Aliases: |
KNTC1AP, hZwilch |
Location: |
Chr.15:66505083-66549485 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZWILCH | NM_001287821.1 | 13 | 1588 | NP_001274750.1 |
| NM_001287822.1 | 12 | 1492 | NP_001274751.1 | |
| NM_001287823.1 | 12 | 1441 | NP_001274752.1 | |
| NM_017975.4 | 13 | 1493 | NP_060445.3 | |
| AK000898.1 | 11 | 1097 | ||
| AK023175.1 | 13 | 1483 | BAB14446.1 | |
| AK027468.1 | 4 | 308 | BAB55133.1 | |
| AK122787.1 | 12 | 1280 | ||
| AK291048.1 | 12 | 1198 | ||
| BC036900.1 | 13 | 1239 | AAH36900.1 | |
| BC090041.1 | 13 | 1337 | AAH90041.1 | |
| BX640701.1 | 13 | 1555 | CAE45821.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581751 | Chr.15:66531168 - 66535962 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map