Assay Details
Target Gene Details
Entrez Gene ID: | 55471 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 7 |
Gene Aliases: |
C2orf56, MidA, PRO1853 |
Location: |
Chr.2:37231631-37259052 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF7 |
Target Gene Details
Entrez Gene ID: | 23683 |
Gene Name: | protein kinase D3 |
Gene Aliases: |
EPK2, PKC-NU, PKD3, PRKCN, nPKC-NU |
Location: |
Chr.2:37250502-37324803 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKD3 | NM_005813.4 | 18 | 3668 | NP_005804.1 |
| XM_005264237.3 | 19 | 3941 | XP_005264294.1 | |
| XM_005264238.3 | 18 | 2917 | XP_005264295.1 | |
| XM_017003741.1 | 20 | 3946 | XP_016859230.1 | |
| XM_017003742.1 | 19 | 2985 | XP_016859231.1 | |
| XM_017003743.1 | 16 | 2667 | XP_016859232.1 | |
| XM_017003744.1 | 15 | 2415 | XP_016859233.1 | |
| AB015982.2 | 18 | 3668 | BAA36514.1 |
More Information
Set Membership: |
|

Genomic Map