Assay Details
Target Gene Details
Entrez Gene ID: | 387357 |
Gene Name: | thymocyte selection associated |
Gene Aliases: |
C6orf190, C6orf207, GASP, SPOT, TSEPA |
Location: |
Chr.6:127697191-127918631 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THEMIS | NM_001010923.2 | 6 | 3500 | NP_001010923.1 |
| NM_001164685.1 | 7 | 3617 | NP_001158157.1 | |
| NM_001164687.1 | 6 | 3464 | NP_001158159.1 | |
| NM_001318531.1 | 7 | 3565 | NP_001305460.1 | |
| XM_011535813.2 | 8 | 3694 | XP_011534115.1 | |
| XM_011535814.2 | 7 | 3465 | XP_011534116.1 | |
| XM_011535816.1 | 8 | 3690 | XP_011534118.1 | |
| XM_017010849.1 | 6 | 3208 | XP_016866338.1 | |
| AK094863.1 | 3 | 1777 | ||
| AK128377.1 | 7 | 3498 |
More Information
Additional Information:
For this assay, SNP(s) [rs113549810] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map