Assay Details
Target Gene Details
Entrez Gene ID: | 6199 |
Gene Name: | ribosomal protein S6 kinase B2 |
Gene Aliases: |
KLS, P70-beta, P70-beta-1, P70-beta-2, S6K-beta2, S6K2, SRK, STK14B, p70(S6K)-beta, p70S6Kb |
Location: |
Chr.11:67428460-67435408 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KB2 | NM_003952.2 | NP_003943.2 | ||
| XM_005274164.1 | XP_005274221.1 | |||
| XM_005274165.4 | XP_005274222.1 | |||
| XM_006718655.3 | XP_006718718.1 | |||
| XM_006718656.3 | XP_006718719.1 | |||
| XM_006718657.1 | XP_006718720.1 | |||
| XM_017018108.1 | XP_016873597.1 | |||
| AB016869.1 | BAA34402.1 | |||
| AB019245.1 | BAA37145.1 | |||
| AB208827.1 | BAD92064.1 | |||
| AF076931.1 | AAD46063.1 | |||
| AF099739.1 | AAD20990.1 | |||
| AK297527.1 | ||||
| BC000094.2 | AAH00094.3 | |||
| BC006106.1 | AAH06106.3 | |||
| BC136564.1 | ||||
| BX337895.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524096 | Chr.11:67418529 - 67457517 on Build GRCh38 | Loss |
|
| dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss |
|
| dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion |
|
| nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map