Assay Details
Target Gene Details
Entrez Gene ID: | 8473 |
Gene Name: | O-linked N-acetylglucosamine (GlcNAc) transferase |
Gene Aliases: |
HINCUT-1, HRNT1, O-GLCNAC |
Location: |
Chr.X:71533062-71575897 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGT | NM_181672.2 | 10 | 1480 | NP_858058.1 |
| NM_181673.2 | 10 | 1450 | NP_858059.1 | |
| XM_017029907.1 | 8 | 1009 | XP_016885396.1 | |
| XM_017029908.1 | 8 | 4482 | XP_016885397.1 | |
| AF223393.1 | 6 | 1117 | AAF31458.1 | |
| AK313530.1 | 6 | 1396 | ||
| AL050366.1 | 5 | 2707 | CAB62528.1 | |
| AL833085.2 | 10 | 1460 | CAD89970.1 | |
| BC014434.1 | 10 | 1381 | AAH14434.1 | |
| BC038180.1 | 10 | 1413 | AAH38180.1 | |
| BX537844.1 | 10 | 1438 | CAD97853.1 | |
| U77413.1 | 6 | 1129 | AAB63466.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3574043 | Chr.X:71548119 - 71559374 on Build GRCh38 | Loss |
|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map