Assay Details
Target Gene Details
Entrez Gene ID: | 11146 |
Gene Name: | glomulin, FKBP associated protein |
Gene Aliases: |
FAP, FAP48, FAP68, FKBPAP, GLML, GVM, VMGLOM |
Location: |
Chr.1:92245786-92441936 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLMN | XM_011540546.2 | XP_011538848.1 |
Target Gene Details
Entrez Gene ID: | 79871 |
Gene Name: | RNA polymerase II associated protein 2 |
Gene Aliases: |
C1orf82, Rtr1 |
Location: |
Chr.1:92298965-92388175 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPAP2 | NM_024813.2 | 8 | 1294 | NP_079089.2 |
| XM_005271223.2 | 8 | 1236 | XP_005271280.1 | |
| XM_011542167.2 | 8 | 1236 | XP_011540469.1 | |
| XM_017002362.1 | 8 | 1236 | XP_016857851.1 | |
| XM_017002363.1 | 8 | 1236 | XP_016857852.1 | |
| XM_017002364.1 | 8 | 1236 | XP_016857853.1 | |
| AK023212.1 | 8 | 1211 | BAB14465.1 | |
| AK225054.1 | 8 | 1206 | ||
| BC031070.1 | 8 | 1200 | AAH31070.1 | |
| BC039014.1 | 8 | 1294 | AAH39014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830592 | Chr.1:92203725 - 92340072 on Build GRCh38 | Gain |
|
| esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map