Assay Details
Target Gene Details
Entrez Gene ID: | 23398 |
Gene Name: | peptidylprolyl isomerase domain and WD repeat containing 1 |
Gene Aliases: |
- |
Location: |
Chr.5:65563236-65587549 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPWD1 | NM_001278926.1 | 6 | 900 | NP_001265855.1 |
| NM_001278927.1 | 5 | 690 | NP_001265856.1 | |
| NM_001278929.1 | 4 | 760 | NP_001265858.1 | |
| NM_015342.3 | 5 | 866 | NP_056157.1 | |
| XM_005248474.3 | XP_005248531.1 | |||
| XM_006714583.3 | 6 | 836 | XP_006714646.1 | |
| XM_011543295.1 | 4 | 1148 | XP_011541597.1 | |
| XM_017009286.1 | XP_016864775.1 | |||
| AK025679.1 | 5 | 813 | ||
| AK298175.1 | 5 | 630 | ||
| AK300022.1 | 6 | 843 | ||
| AK301654.1 | 4 | 700 | ||
| BC015385.1 | 5 | 794 | AAH15385.1 | |
| D38552.1 | 5 | 789 | BAA07555.1 |
More Information
Set Membership: |
|

Genomic Map