Assay Details
Target Gene Details
Entrez Gene ID: | 5653 |
Gene Name: | kallikrein related peptidase 6 |
Gene Aliases: |
Bssp, Klk7, PRSS18, PRSS9, SP59, hK6 |
Location: |
Chr.19:50958631-50969673 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK6 | NM_001012964.2 | 4 | 709 | NP_001012982.1 |
| NM_001012965.2 | 3 | 661 | NP_001012983.1 | |
| NM_001319948.1 | 4 | 355 | NP_001306877.1 | |
| NM_001319949.1 | 3 | 552 | NP_001306878.1 | |
| NM_002774.3 | 5 | 512 | NP_002765.1 | |
| AF013988.1 | 5 | 414 | AAB66483.1 | |
| AK314897.1 | 5 | 417 | ||
| AY279383.1 | 4 | 356 | ||
| AY318867.1 | 4 | 518 | AAP82446.1 | |
| AY318868.1 | 3 | 347 | ||
| AY318869.1 | 4 | 504 | AAP82448.1 | |
| AY318870.1 | 3 | 456 | ||
| AY457039.1 | AAR23148.1 | |||
| BC015525.2 | 4 | 485 | AAH15525.1 | |
| BT006852.1 | 3 | 268 | AAP35498.1 | |
| D78203.1 | 5 | 423 | BAA11306.1 | |
| DQ223012.1 | 3 | 360 | ABB04464.1 | |
| U62801.1 | 5 | 513 | AAB07113.1 |
More Information
Set Membership: |
|

Genomic Map