Assay Details
Target Gene Details
Entrez Gene ID: | 1831 |
Gene Name: | TSC22 domain family member 3 |
Gene Aliases: |
DIP, DSIPI, GILZ, TSC-22R |
Location: |
Chr.X:107713221-107777329 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
More Information
Set Membership: |
|

Genomic Map