Assay Details
Target Gene Details
Entrez Gene ID: | 51742 |
Gene Name: | AT-rich interaction domain 4B |
Gene Aliases: |
BCAA, BRCAA1, RBBP1L1, RBP1L1, SAP180 |
Location: |
Chr.1:235166895-235328240 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID4B | NM_001206794.1 | 20 | 2680 | NP_001193723.1 |
| NM_016374.5 | 20 | 2801 | NP_057458.4 | |
| NM_031371.3 | 19 | 2543 | NP_112739.2 | |
| NR_135074.1 | 20 | 2803 | ||
| XM_006711781.2 | 20 | 2716 | XP_006711844.1 | |
| XM_011544212.2 | 20 | 2640 | XP_011542514.1 | |
| XM_017001468.1 | 19 | 2462 | XP_016856957.1 | |
| XM_017001469.1 | 19 | 2355 | XP_016856958.1 | |
| XM_017001470.1 | 18 | 2203 | XP_016856959.1 | |
| XM_017001471.1 | 18 | 2207 | XP_016856960.1 | |
| XM_017001472.1 | 10 | 1474 | XP_016856961.1 | |
| AB210032.1 | 20 | 2680 | ||
| AF083249.1 | 4 | 667 | AAD41239.1 | |
| AF208045.2 | 18 | 2104 | AAF23433.3 | |
| AF214114.2 | 19 | 2509 | AAF28341.2 | |
| AF227899.1 | 6 | 986 | AAF36964.1 | |
| AY220790.1 | 18 | 2303 | AAO63590.1 | |
| BC130418.1 | 20 | 2718 | ||
| BX648820.1 | 19 | 2495 | CAI46047.1 |
More Information
Set Membership: |
|

Genomic Map