Assay Details
Target Gene Details
Entrez Gene ID: | 121457 |
Gene Name: | IKBKB interacting protein |
Gene Aliases: |
IKIP |
Location: |
Chr.12:98613404-98645051 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKBIP | NM_201612.2 | 3 | 1257 | NP_963906.1 |
| NM_201613.2 | 2 | 1139 | NP_963907.1 | |
| AJ539426.1 | 3 | 1017 | CAD62381.1 | |
| AJ539427.1 | 2 | 899 | CAD62382.1 | |
| AK123069.1 | 2 | 1043 | BAC85528.1 | |
| BC029415.1 | 2 | 899 | ||
| BC051372.1 | 2 | 868 | ||
| BC107572.1 | 2 | 660 | AAI07573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559918 | Chr.12:98570959 - 98666572 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map