Assay Details
Target Gene Details
Entrez Gene ID: | 54457 |
Gene Name: | TATA-box binding protein associated factor 7 like |
Gene Aliases: |
CT40, TAF2Q |
Location: |
Chr.X:101268253-101293071 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF7L | NM_001168474.1 | NP_001161946.1 | ||
| NM_024885.3 | NP_079161.3 | |||
| XM_005262152.1 | XP_005262209.1 | |||
| XM_006724664.1 | XP_006724727.1 | |||
| AF285595.1 | AAK31974.1 | |||
| AK026810.1 | 1 | 270 | BAB15560.1 | |
| AK308261.1 | ||||
| BC043391.1 | AAH43391.1 | |||
| DB042169.1 | ||||
| DB455298.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524411 | Chr.X:101190975 - 101377649 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map