Assay Details
Target Gene Details
Entrez Gene ID: | 10231 |
Gene Name: | regulator of calcineurin 2 |
Gene Aliases: |
CSP2, DSCR1L1, MCIP2, RCN2, ZAKI-4, ZAKI4 |
Location: |
Chr.6:46220730-46492067 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCAN2 | NM_001251973.1 | 5 | 2372 | NP_001238902.1 |
| NM_001251974.1 | 5 | 2325 | NP_001238903.1 | |
| NM_005822.3 | 4 | 2447 | NP_005813.2 | |
| XM_011514226.1 | 5 | 2414 | XP_011512528.1 | |
| AK090990.1 | 5 | 2296 | ||
| AY034085.1 | 5 | 2279 | AAK59805.1 | |
| AY034086.1 | 5 | 2292 | AAK59806.1 | |
| BC038509.1 | 4 | 2230 | AAH38509.1 | |
| BP264912.1 | 1 | 86 | ||
| D83407.1 | 4 | 2223 | BAA11911.1 |
More Information
Set Membership: |
|

Genomic Map