Assay Details
Target Gene Details
Entrez Gene ID: | 29883 |
Gene Name: | CCR4-NOT transcription complex subunit 7 |
Gene Aliases: |
CAF-1, CAF1, Caf1a, hCAF-1 |
Location: |
Chr.8:17226030-17246909 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT7 | NM_001322090.1 | 8 | 1859 | NP_001309019.1 |
| NM_001322091.1 | 7 | 1644 | NP_001309020.1 | |
| NM_001322092.1 | 8 | 1719 | NP_001309021.1 | |
| NM_001322093.1 | 8 | 1850 | NP_001309022.1 | |
| NM_001322094.1 | NP_001309023.1 | |||
| NM_001322095.1 | 8 | 1888 | NP_001309024.1 | |
| NM_001322096.1 | 8 | 1965 | NP_001309025.1 | |
| NM_001322097.1 | 8 | 1825 | NP_001309026.1 | |
| NM_013354.6 | 7 | 1784 | NP_037486.2 | |
| XM_005273481.2 | 8 | 1890 | XP_005273538.1 | |
| AL528251.3 | ||||
| AL833642.1 | 7 | 1623 | ||
| BC007315.2 | 7 | 1598 | AAH07315.1 | |
| BC060852.1 | 7 | 1744 | AAH60852.1 | |
| BM468592.1 | 1 | 465 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831244 | Chr.8:17105917 - 17270710 on Build GRCh38 | Gain |
|
| nsv981916 | Chr.8:17229359 - 17230462 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map