Assay Details
Target Gene Details
Entrez Gene ID: | 164 |
Gene Name: | adaptor related protein complex 1 gamma 1 subunit |
Gene Aliases: |
ADTG, CLAPG1 |
Location: |
Chr.16:71729000-71809073 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1G1 | NM_001030007.1 | 24 | 3046 | NP_001025178.1 |
| NM_001128.5 | 23 | 3037 | NP_001119.3 | |
| AB015317.1 | 23 | 2742 | BAA33389.1 | |
| AK055122.1 | 7 | 1657 | ||
| AK128078.1 | 26 | 3592 | ||
| AK293233.1 | 15 | 1661 | ||
| AL110198.1 | 7 | 1111 | CAB53673.1 | |
| BC036283.1 | 24 | 3011 | AAH36283.1 | |
| Y12226.1 | 24 | 2760 | CAA72902.1 |
More Information
Set Membership: |
|

Genomic Map