Assay Details
Target Gene Details
Entrez Gene ID: | 341 |
Gene Name: | apolipoprotein C1 |
Gene Aliases: |
Apo-CI, ApoC-I, apo-CIB, apoC-IB |
Location: |
Chr.19:44914247-44919349 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOC1 | NM_001321065.1 | 3 | 387 | NP_001307994.1 |
| NM_001321066.1 | 4 | 449 | NP_001307995.1 | |
| NM_001645.4 | 3 | 323 | NP_001636.1 | |
| AI310334.1 | 2 | 251 | ||
| AI885116.1 | 2 | 241 | ||
| AK225971.1 | 3 | 309 | ||
| AK312036.1 | 3 | 204 | ||
| BC009698.2 | 2 | 146 | AAH09698.1 | |
| BC055093.1 | 4 | 375 | AAH55093.1 | |
| BT007142.1 | 2 | 124 | AAP35806.1 | |
| CA445083.1 | 2 | 266 | ||
| CR456907.1 | 2 | 124 | CAG33188.1 | |
| DA482306.1 | 4 | 449 | ||
| M27359.1 | 2 | 82 | AAA51762.1 | |
| X00570.1 | 2 | 180 | CAA25235.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv138296 | Chr.19:44915738 - 44918040 on Build GRCh38 | Deletion |
|
| nsv509747 | Chr.19:44877936 - 44943426 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map