Assay Details
Target Gene Details
Entrez Gene ID: | 4632 |
Gene Name: | myosin light chain 1 |
Gene Aliases: |
MLC1F, MLC3F |
Location: |
Chr.2:210290144-210315171 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYL1 | NM_079420.2 | NP_524144.1 | ||
| NM_079422.2 | NP_524146.1 | |||
| AK311892.1 | ||||
| AK311942.1 | ||||
| BC005318.1 | AAH05318.1 | |||
| CR456869.1 | CAG33150.1 | |||
| DA898482.1 | ||||
| M20643.1 | AAA59855.1 | |||
| X05451.1 | CAA29020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012567 | Chr.2:210222864 - 210450830 on Build GRCh38 | Gain |
|
| dgv4163n100 | Chr.2:210278131 - 210350344 on Build GRCh38 | Gain |
|
| nsv834520 | Chr.2:210236541 - 210392344 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map