Assay Details
Target Gene Details
Entrez Gene ID: | 169792 |
Gene Name: | GLIS family zinc finger 3 |
Gene Aliases: |
NDH, ZNF515 |
Location: |
Chr.9:3824127-4310694 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLIS3 | NM_001042413.1 | 11 | 7319 | NP_001035878.1 |
| NM_152629.3 | 10 | 6335 | NP_689842.3 | |
| XM_005251386.4 | 10 | 6878 | XP_005251443.1 | |
| XM_005251387.4 | 10 | 7156 | XP_005251444.1 | |
| XM_005251388.4 | 9 | 6670 | XP_005251445.1 | |
| XM_011517763.2 | 11 | 6998 | XP_011516065.1 | |
| XM_011517764.2 | 11 | 6828 | XP_011516066.1 | |
| XM_011517766.2 | 10 | 7407 | XP_011516068.1 | |
| XM_011517767.2 | 10 | 6801 | XP_011516069.1 | |
| XM_017014361.1 | 10 | 7106 | XP_016869850.1 | |
| AA933816.1 | ||||
| AB209404.1 | 8 | 4928 | BAD92641.1 | |
| AK075059.1 | 3 | 2074 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1028973 | Chr.9:3789264 - 3856678 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map