Assay Details
Target Gene Details
Entrez Gene ID: | 27293 |
Gene Name: | sphingomyelin phosphodiesterase acid like 3B |
Gene Aliases: |
ASML3B |
Location: |
Chr.1:27934955-27959157 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMPDL3B | NM_001304579.1 | 8 | 1509 | NP_001291508.1 |
| NM_014474.3 | 8 | 1438 | NP_055289.2 | |
| XM_011541259.2 | 9 | 1528 | XP_011539561.1 | |
| XM_011541260.2 | 9 | 2163 | XP_011539562.1 | |
| AK293489.1 | 8 | 1062 | ||
| AK293566.1 | 8 | 1435 | ||
| AK301349.1 | 8 | 1393 | ||
| AK316500.1 | 8 | 1364 | ||
| BQ011468.1 | ||||
| BX389470.1 | 4 | 571 | ||
| CA425405.1 | 1 | 497 | ||
| Y08134.1 | 8 | 1330 | CAA69328.1 |
More Information
Set Membership: |
|

Genomic Map