Assay Details
Target Gene Details
Entrez Gene ID: | 55858 |
Gene Name: | transmembrane protein 165 |
Gene Aliases: |
CDG2K, FT27, GDT1, TMPT27, TPARL |
Location: |
Chr.4:55395913-55453397 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM165 | NM_018475.4 | 2 | 526 | NP_060945.2 |
| NR_073070.1 | 3 | 629 | ||
| XM_011534394.2 | 2 | 516 | XP_011532696.1 | |
| XM_017008412.1 | 4 | 444 | XP_016863901.1 | |
| AF132746.1 | 2 | 249 | AAL75947.1 | |
| AF183409.1 | 2 | 249 | AAG09678.1 | |
| AF220188.1 | 2 | 443 | AAF67653.1 | |
| AK290663.1 | 2 | 479 | ||
| AK295289.1 | 3 | 579 | ||
| BC104978.1 | 2 | 324 | AAI04979.1 | |
| BC104980.1 | 2 | 324 | AAI04981.1 | |
| BC107582.1 | 2 | 486 | AAI07583.1 | |
| DA371259.1 | ||||
| HY089602.1 |
More Information
Set Membership: |
|

Genomic Map