Assay Details
Target Gene Details
Entrez Gene ID: | 1499 |
Gene Name: | catenin beta 1 |
Gene Aliases: |
CTNNB, MRD19, armadillo |
Location: |
Chr.3:41199451-41240448 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTNNB1 | NM_001098209.1 | NP_001091679.1 | ||
| NM_001098210.1 | NP_001091680.1 | |||
| NM_001904.3 | NP_001895.1 | |||
| XM_005264886.2 | XP_005264943.1 | |||
| XM_006712983.1 | XP_006713046.1 | |||
| XM_006712984.1 | XP_006713047.1 | |||
| XM_006712985.1 | XP_006713048.1 | |||
| XM_017005738.1 | XP_016861227.1 | |||
| AB062292.1 | BAB93475.1 | |||
| AB451264.1 | ||||
| AB451392.1 | ||||
| AK289932.1 | ||||
| AK294778.1 | ||||
| AK296790.1 | ||||
| AK299954.1 | ||||
| AK314776.1 | ||||
| BC058926.1 | AAH58926.1 | |||
| X87838.1 | CAA61107.1 | |||
| Z19054.1 | CAA79497.1 |
More Information
Set Membership: |
|

Genomic Map